Ethyl (methylthio)acetate ≥98.0%
Leverantör: TCI
|
Varning
|
M0897-25G
M0897-5G
TCIAM0897-25GEA
565
SEK
TCIAM0897-25G
TCIAM0897-5G
Ethyl (methylthio)acetate ≥98.0%
Ethyl (methylthio)acetate
Formel:
C₅H₁₀O₂S Kokpunkt: 176°C Densitet: 1,04 g/cm³ (20 °C) Flampunkt: 59°C |
MDL Number:
MFCD00009182 CAS nummer: 4455-13-4 EINECS: 224-700-2 UN: 3272 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...