(2-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Ethyl 2-Pyridylacetate, Ethyl 2-pyridineacetate, Ethyl (2-pyridyl)acetate
P1075-25G P1075-5G
TCIAP1075-25GEA 2100 SEK
TCIAP1075-25G TCIAP1075-5G
(2-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
(2-Pyridyl)acetic acid ethyl ester
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
Kokpunkt: 122°C
Smältpunkt: 86…90 °C
Densitet: 1,08 g/cm³ (20 °C)
MDL Number: MFCD00006359
CAS nummer: 2739-98-2
EINECS: 220-365-1
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %