Prednisolone ≥98.0%
Leverantör: TCI
|
Varning
|
P0637-1G
P0637-5G
P0637-25G
TCIAP0637-1GEA
455
SEK
TCIAP0637-1G
TCIAP0637-5G
TCIAP0637-25G
Prednisolone ≥98.0%
Prednisolone
Formel:
C₂₁H₂₈O₅ Kokpunkt: 571 °C (1013 hPa) Smältpunkt: 246 °C Densitet: 1,212 g/cm³ (20 °C) |
MDL Number:
MFCD00003649 CAS nummer: 50-24-8 EINECS: 200-021-7 Merck Index: 14,07721 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | +97.0 to +103.0 deg (C=1, Dioxane) |
Solubility in Dioxane | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...