2-Acetyl-1-naphthol ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1'-Hydroxy-2'-acetonaphthone, 1-(1-Hydroxynaphthalen-2-yl)ethan-1-one, 2-Acetyl-1-hydroxynaphthalene, 1-Hydroxy-2-acetonaphthone
H0935-100G H0935-25G
TCIAH0935-100GEA 6850 SEK
TCIAH0935-100G TCIAH0935-25G
2-Acetyl-1-naphthol ≥98.0%
2-Acetyl-1-naphthol
Formel: C₁₂H₁₀O₂
MW: 186,21 g/mol
Kokpunkt: 110 - 115 °C (0,5 -rr)
Smältpunkt: 325 °C
MDL Number: MFCD00003963
CAS nummer: 711-79-5
EINECS: 211-918-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c9fca17b27f8d334c7238d6976a63df5c237590&vid=5c9fca17b27f8d334c7238d6976a63df5c237590&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 100.0 deg-C
Solubility in hot Methanol within almost transparency