7-Hydroxyflavone ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Hydroxy-2-phenylchromen-4-one, 7-Hydroxy-2-phenyl-4-benzopyrone, 7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, 7-Hydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 7-Hydroxy-2-phenyl-4H-chromen-4-one

H0852-1G H0852-5G
TCIAH0852-1GEA 455 SEK
TCIAH0852-1G TCIAH0852-5G
7-Hydroxyflavone ≥97.0%
7-Hydroxyflavone
Formel: C₁₅H₁₀O₃
MW: 238,24 g/mol
Kokpunkt: 395 °C (1013 hPa)
Smältpunkt: 244…244,5 °C
MDL Number: MFCD00006835
CAS nummer: 6665-86-7
EINECS: 229-705-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d05d24e7a8c650236db9459b4a4ceb5bfec4b06&vid=0d05d24e7a8c650236db9459b4a4ceb5bfec4b06&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR