2'-Hydroxy-5'-methylacetophenone ≥98.0%
Leverantör: TCI
Varning
H0790-25G H0790-5G
TCIAH0790-25GEA 1275 SEK
TCIAH0790-25G TCIAH0790-5G
2'-Hydroxy-5'-methylacetophenone ≥98.0%
2'-Hydroxy-5'-methylacetophenone
Formel: C₉H₁₀O₂
Kokpunkt: 83 - 87 °C (0,5 -rr)
Smältpunkt: 53 °C
Densitet: 1,0797 g/cm³ (53 °C)
MDL Number: MFCD00002380
CAS nummer: 1450-72-2
EINECS: 215-915-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a4a6e6c7e7faaf5cf0e7fc53fe4fc60c662caa&vid=62a4a6e6c7e7faaf5cf0e7fc53fe4fc60c662caa&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C
Solubility in Methanol within almost transparency