6-Methoxyflavone ≥99.0%

Leverantör: TCI EUROPE

Synonyms: 6-Methoxy-2-phenyl-4H-1-benzopyran-4-one, 6-Methoxy-2-phenyl-4H-benzo[b]pyran-4-one, 6-Methoxy-2-phenyl-4-benzopyrone, 6-Methoxy-2-phenyl-4H-chromen-4-one, 6-Methoxy-2-phenylchromen-4-one

M1346-1G M1346-5G
TCIAM1346-1GEA 405 SEK
TCIAM1346-1G TCIAM1346-5G
6-Methoxyflavone ≥99.0%
6-Methoxyflavone
Formel: C₁₆H₁₂O₃
MW: 252,27 g/mol
MDL Number: MFCD00017322
CAS nummer: 26964-24-9
EINECS: 248-144-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=093c865b7324d4d5e4fedac2194d071d38b92cb8&vid=093c865b7324d4d5e4fedac2194d071d38b92cb8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 162.0 to 166.0 deg-C
Solubility in hot DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR