Norcamphor ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone

N0511-25G N0511-5G
TCIAN0511-25GEA 3950 SEK
TCIAN0511-25G TCIAN0511-5G
Norcamphor ≥98.0%
Norcamphor
Formel: C₇H₁₀O
MW: 110,16 g/mol
Kokpunkt: 55°C
Smältpunkt: –88 °C
Densitet: 1,03 g/cm³ (25 °C)
MDL Number: MFCD00074823
CAS nummer: 497-38-1
EINECS: 207-846-1
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b52e990ef37a1b1d25b3b948054ec5089738e954&vid=b52e990ef37a1b1d25b3b948054ec5089738e954&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR