2-Acetylfuran ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Furyl methyl ketone, 1-(Furan-2-yl)ethanone

A0091-25G A0091-500G
TCIAA0091-25GEA 525 SEK
TCIAA0091-25G TCIAA0091-500G
2-Acetylfuran ≥98.0%
2-Acetylfuran
Formel: C₆H₆O₂
MW: 110,11 g/mol
Kokpunkt: 173°C
Smältpunkt: 29…33 °C
Densitet: 1,098 g/cm³ (20 °C)
MDL Number: MFCD00003242
CAS nummer: 1192-62-7
EINECS: 214-757-1
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=881b70bfce925a7141a613a01627c72adad6ae05&vid=881b70bfce925a7141a613a01627c72adad6ae05&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR