Flavone ≥98.0%

Leverantör: TCI

F0015-1G F0015-5G
TCIAF0015-1GEA 215 SEK
TCIAF0015-1G TCIAF0015-5G
Flavone ≥98.0%
Flavone
Formel: C₁₅H₁₀O₂
Kokpunkt: 185°C
Smältpunkt: 96 °C
MDL Number: MFCD00006825
CAS nummer: 525-82-6
EINECS: 208-383-8
Merck Index: 14,04092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0c3c725008417ce95fb91c99a72597d6dc259b&vid=5f0c3c725008417ce95fb91c99a72597d6dc259b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 98.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR