4'-Octylacetophenone ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 1-(4-Octylphenyl)ethanone, p-Octylacetophenone

O0207-25G O0207-5G
TCIAO0207-25GEA 3600 SEK
TCIAO0207-25G TCIAO0207-5G
4'-Octylacetophenone ≥97.0%
4'-Octylacetophenone
Formel: C₁₆H₂₄O
MW: 232,37 g/mol
MDL Number: MFCD00043682
CAS nummer: 10541-56-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c176efa353d2996364fc490cc9d4c457f6aa2f7&vid=6c176efa353d2996364fc490cc9d4c457f6aa2f7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow to Green powder to lump to clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR