Diaminomaleonitrile ≥96.0%
Leverantör: TCI EUROPE
|
Fara
|
D1335-25G
D1335-500G
D1335-100G
TCIAD1335-25GEA
365
SEK
TCIAD1335-25G
TCIAD1335-500G
TCIAD1335-100G
Diaminomaleonitrile ≥96.0%
Diaminomaleonitrile
Formel:
C₄H₄N₄ MW: 108,1 g/mol Kokpunkt: 445 °C (1013 hPa) Smältpunkt: 178…181 °C Densitet: 1,368 g/cm³ (20 °C) |
MDL Number:
MFCD00001870 CAS nummer: 1187-42-4 EINECS: 214-697-6 UN: 3439 ADR: 6.1,III |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...