4-Chlorobutyl acetate ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Acetic acid 4-chlorobutyl ester
A1098-25G A1098-250G
TCIAA1098-25GEA 485 SEK
TCIAA1098-25G TCIAA1098-250G
4-Chlorobutyl acetate ≥97.0%
4-Chlorobutyl acetate
Formel: C₆H₁₁ClO₂
MW: 150,6 g/mol
Kokpunkt: 87°C
Densitet: 1,1024 g/cm³ (0 °C)
Flampunkt: 64°C
MDL Number: MFCD00001013
CAS nummer: 6962-92-1
EINECS: 230-158-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7867f23316f33509f5e78a649512306343c46710&vid=7867f23316f33509f5e78a649512306343c46710&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %