3-Chloro-4-hydroxybenzaldehyde ≥97.0% (by GC, titration analysis)

Leverantör: TCI
Varning

C2183-25G C2183-5G
TCIAC2183-25GEA 13350 SEK
TCIAC2183-25G TCIAC2183-5G
3-Chloro-4-hydroxybenzaldehyde ≥97.0% (by GC, titration analysis)
3-Chloro-4-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
Kokpunkt: 150°C
Smältpunkt: 133…134 °C
Densitet: 1,0404 g/cm³ (20 °C)
MDL Number: MFCD00016981
CAS nummer: 2420-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7360c19003c5d4a3e1d0f8db0039117d5ad8a486&vid=7360c19003c5d4a3e1d0f8db0039117d5ad8a486&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR