4-Bromo-2-fluorobenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Brom-4-cyan-3-fluorbenzene

B2745-25G B2745-5G
TCIAB2745-25GEA 1875 SEK
TCIAB2745-25G TCIAB2745-5G
4-Bromo-2-fluorobenzonitrile ≥98.0%
4-Bromo-2-fluorobenzonitrile
Formel: C₇H₃BrFN
MW: 200,01 g/mol
Kokpunkt: 238 - 239 °C (1013 hPa)
Smältpunkt: 71…73 °C
MDL Number: MFCD00143264
CAS nummer: 105942-08-3
UN: 3439
ADR: 6.1,III

Order Now

Specifikation testresultat

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR