(4-Bromophenyl)acetic acid ethyl ester ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate
E0903-25G
E0903-5G
TCIAE0903-25GEA
1150
SEK
TCIAE0903-25G
TCIAE0903-5G
(4-Bromophenyl)acetic acid ethyl ester ≥98.0%
(4-Bromophenyl)acetic acid ethyl ester
Formel:
C₁₀H₁₁BrO₂ MW: 243,1 g/mol Kokpunkt: 144°C |
MDL Number:
MFCD00016333 CAS nummer: 14062-25-0 |
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...