2-Bromoethyl acetate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Acetic acid (2-bromoethyl) ester
A0023-25G A0023-500G
TCIAA0023-25GEA 310 SEK
TCIAA0023-25G TCIAA0023-500G
2-Bromoethyl acetate ≥98.0%
2-Bromoethyl acetate
Formel: C₄H₇BrO₂
MW: 167 g/mol
Kokpunkt: 162°C
Smältpunkt: –14 °C
Densitet: 1,4623 g/cm³ (20 °C)
Flampunkt: 71°C
MDL Number: MFCD00000235
CAS nummer: 927-68-4
EINECS: 213-159-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6772feda2508068160175edb3d3349f91a20131a&vid=6772feda2508068160175edb3d3349f91a20131a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %