5-Bromo-2-methoxybenzonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

B3123-5G B3123-25G
TCIAB3123-5GEA 1075 SEK
TCIAB3123-5G TCIAB3123-25G
5-Bromo-2-methoxybenzonitrile ≥98.0%
5-Bromo-2-methoxybenzonitrile
Formel: C₈H₆BrNO
MW: 212,05 g/mol
MDL Number: MFCD00143429
CAS nummer: 144649-99-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cf684d9af387c5ec3df98af7d3b2fb4f9e500cc&vid=0cf684d9af387c5ec3df98af7d3b2fb4f9e500cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR