2-Bromo-4-chloro-1-fluorobenzene 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene

B2956-25G B2956-5G
TCIAB2956-25GEA 1350 SEK
TCIAB2956-25G TCIAB2956-5G
2-Bromo-4-chloro-1-fluorobenzene 98,0 GC_ANALYSMETOD
2-Bromo-4-chloro-1-fluorobenzene
Formel: C₆H₃BrClF
MW: 209.44 g/mol
Kokpunkt: 90°C
Flampunkt: 85°C
MDL Number: MFCD00672934
CAS nummer: 1996-30-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b32da8c5025beff9b75b4feb05638dba9698f1f3&vid=b32da8c5025beff9b75b4feb05638dba9698f1f3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR