3-Bromo-5-nitrosalicylaldehyde ≥95.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Bromo-2-hydroxy-5-nitrobenzaldehyde
B3198-1G
B3198-5G
TCIAB3198-1GEA
470
SEK
TCIAB3198-1G
TCIAB3198-5G
3-Bromo-5-nitrosalicylaldehyde ≥95.0% (by GC, titration analysis)
3-Bromo-5-nitrosalicylaldehyde
Formel:
C₇H₄BrNO₄ MW: 246,02 g/mol |
MDL Number:
MFCD00051833 CAS nummer: 16789-84-7 UN: 2811 ADR: 6.1,III |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 146.0 to 151.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...