Sodium triacetoxyborohydride ≥80.0%
Leverantör: TCI
|
Varning
|
S0394-100G
S0394-25G
TCIAS0394-100GEA
2050
SEK
TCIAS0394-100G
TCIAS0394-25G
Sodium triacetoxyborohydride ≥80.0%
Sodium triacetoxyborohydride
Formel:
C₆H₁₀BNaO₆ MW: 211.94 g/mol |
MDL Number:
MFCD00012211 CAS nummer: 56553-60-7 UN: 2813 ADR: 4.3 III Merck Index: 14,08695 |
|
Specifikation testresultat
Appearance | White powder to crystal |
Purity (NBS Method) | min. 80.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...