Propionaldehyd ≥98.0%
Leverantör: TCI
|
Fara
|
P0498-25ML
P0498-500ML
TCIAP0498-25MLEA
215
SEK
TCIAP0498-25ML
TCIAP0498-500ML
Propionaldehyd ≥98.0%
Propionaldehyd
Formel:
C₃H₆O Kokpunkt: 46°C Smältpunkt: -81°C Densitet: 0,811 g/cm³ (20 °C) Flampunkt: -37°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007020 CAS nummer: 123-38-6 EINECS: 204-623-0 UN: 1275 ADR: 3,II Merck Index: 14,07823 |
Specifikation testresultat
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...