1-Bromo-5-chloropentane ≥98.0%

Leverantör: TCI

B1476-25G B1476-5G
TCIAB1476-25GEA 725 SEK
TCIAB1476-25G TCIAB1476-5G
1-Bromo-5-chloropentane ≥98.0%
1-Bromo-5-chloropentane
Formel: BrH₂C(CH₂)₃CH₂Cl
Kokpunkt: 212°C
Densitet: 1,408 g/cm³ (20 °C)
Flampunkt: 95°C
Storage Temperature: Omgivande
MDL Number: MFCD00001016
CAS nummer: 54512-75-3
EINECS: 259-193-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29f75c0a5352793b8720c5cd7bd50bc563f938a&vid=d29f75c0a5352793b8720c5cd7bd50bc563f938a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR