1-Bromo-3-chloropropane ≥99.0%

Leverantör: TCI EUROPE
Varning

B0575-25G B0575-500G
TCIAB0575-25GEA 215 SEK
TCIAB0575-25G TCIAB0575-500G
1-Bromo-3-chloropropane ≥99.0%
1-Bromo-3-chloropropane
Formel: BrH₂CCH₂CH₂Cl
MW: 157,44 g/mol
Kokpunkt: 143°C
Smältpunkt: -59°C
Densitet: 1,6 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000998
CAS nummer: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba260df4e3c1b073bf5525c1f3b5fd75974ce8d9&vid=1086ee8a53a06dd33de660d3ca4441228c1bc93b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR