Tetrabromomethane ≥99.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Carbon tetrabromide
T0038-25G T0038-500G T0038-100G
TCIAT0038-25GEA 230 SEK
TCIAT0038-25G TCIAT0038-500G TCIAT0038-100G
Tetrabromomethane ≥99.0%
Tetrabromomethane
Formel: Br₄C
MW: 331,63 g/mol
Kokpunkt: 190°C
Smältpunkt: 90…92 °C
Densitet: 3,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000117
CAS nummer: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62de0aa4386510c11d0f23123e50ff34c416b887&vid=bc2f8ab2ddcde000cb3931c84382697e4903a2ad&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White powder to crystal
Purity (GC) min. 99.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in Methanol almost transparency