1-Bromoundecane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Undecyl bromide

B0934-25ML B0934-500ML
TCIAB0934-25MLEA 415 SEK
TCIAB0934-25ML TCIAB0934-500ML
1-Bromoundecane ≥98.0%
1-Bromoundecane
Formel: BrH₂C(CH₂)₉CH₃
MW: 235,21 g/mol
Kokpunkt: 138°C
Smältpunkt: -9°C
Densitet: 1,05 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000223
CAS nummer: 693-67-4
EINECS: 211-757-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db84c99fa1d932e1a6fc9e80d4df68c95a04952e&vid=f22274209caad5f1fb72110832c34008c1702ea8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR