1-Bromopentadecane ≥98.0%
Leverantör: TCI EUROPE
B0936-25ML
B0936-250ML
TCIAB0936-25MLEA
1150
SEK
TCIAB0936-25ML
TCIAB0936-250ML
1-Bromopentadecane ≥98.0%
1-Bromopentadecane
Formel:
BrH₂C(CH₂)₁₃CH₃ MW: 291,32 g/mol Kokpunkt: 158°C Smältpunkt: 19 °C Densitet: 1,0052 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00000229 CAS nummer: 629-72-1 EINECS: 211-104-2 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...