1-Bromohexadecane ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
B0599-25G
B0599-500G
B0599-100G
TCIAB0599-25GEA
215
SEK
TCIAB0599-25G
TCIAB0599-500G
TCIAB0599-100G
1-Bromohexadecane ≥96.0%
1-Bromohexadecane
Formel:
BrH₂C(CH₂)₁₄CH₃ MW: 305,34 g/mol Kokpunkt: 336 °C (1013 hPa) Smältpunkt: 14 °C Densitet: 0,9992 g/cm³ (20 °C) Flampunkt: 181°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00000230 CAS nummer: 112-82-3 EINECS: 204-008-7 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...