1,2,3-Tribromopropane ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
T0355-25G
T0355-500G
TCIAT0355-25GEA
270
SEK
TCIAT0355-25G
TCIAT0355-500G
1,2,3-Tribromopropane ≥97.0%
1,2,3-Tribromopropane
Formel:
BrH₂CBrCHCH₂Br MW: 280,78 g/mol Kokpunkt: 220°C Smältpunkt: 16 °C Densitet: 2,43 g/cm³ (0 °C) Flampunkt: 93°C |
MDL Number:
MFCD00017884 CAS nummer: 96-11-7 EINECS: 202-478-8 Merck Index: 14,09614 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...