9,10-Dihydroanthracene ≥98.0%
Leverantör: TCI
|
Varning
|
D0549-25G
D0549-100G
TCIAD0549-25GEA
815
SEK
TCIAD0549-25G
TCIAD0549-100G
9,10-Dihydroanthracene ≥98.0%
9,10-Dihydroanthracene
Formel:
C₁₄H₁₂ Kokpunkt: 313°C Smältpunkt: 70…72 °C Densitet: 1,207 g/cm³ (20 °C) |
MDL Number:
MFCD00001239 CAS nummer: 613-31-0 EINECS: 210-336-1 UN: 3077 ADR: 9,III |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 108.0 to 111.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...