(±)-1-Indanol ≥99.0%
Leverantör: TCI
Varning
Synonyms: (±)-1-Hydroxyindan, 1-Indanol, 2,3-Dihydro-1H-inden-1-ol
H0516-25G H0516-5G
TCIAH0516-25GEA 935 SEK
TCIAH0516-25G TCIAH0516-5G
(±)-1-Indanol ≥99.0%
(±)-1-Indanol
Formel: C₉H₁₀O
MW: 134.18 g/mol
Kokpunkt: 220°C
Storage Temperature: Kylskåp
MDL Number: MFCD00003797
CAS nummer: 6351-10-6
EINECS: 228-755-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b838007a37cbc281dcbb1a0a2fc24b888351b74d&vid=20f58553421f33b21aa5b1c38f5a313cfe87298f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 52.0 to 55.0 deg-C
Solubility in Methanol very faint turbidity