(±)-3-Octanol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol

O0121-25ML O0121-500ML O0121-100ML
TCIAO0121-25MLEA 470 SEK
TCIAO0121-25ML TCIAO0121-500ML TCIAO0121-100ML
(±)-3-Octanol ≥98.0%
(±)-3-Octanol
Formel: C₈H₁₈O
MW: 130,23 g/mol
Kokpunkt: 177°C
Smältpunkt: –45 °C
Densitet: 0,819 g/cm³ (20 °C)
Flampunkt: 68°C
Storage Temperature: Omgivande
MDL Number: MFCD00004590
CAS nummer: 589-98-0
EINECS: 209-667-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53306b86c8ffe59cb4402885bc181460869a35b0&vid=da1db8c476292949544e34b4ca69307f2dd0459f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR