4,4'-Dimethylbenzophenone ≥99.0%

Leverantör: TCI

D0685-25G D0685-5G
TCIAD0685-25GEA 1100 SEK
TCIAD0685-25G TCIAD0685-5G
4,4'-Dimethylbenzophenone ≥99.0%
4,4'-Dimethylbenzophenone
Formel: C₁₅H₁₄O
Kokpunkt: 151°C
Smältpunkt: 92 °C
Densitet: 1,18 g/cm³ (20 °C)
MDL Number: MFCD00017214
CAS nummer: 611-97-2
EINECS: 210-287-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b433b564fce1d8e7c767263d3cc7a6ee3ced79c9&vid=b433b564fce1d8e7c767263d3cc7a6ee3ced79c9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 92.0 to 95.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR