(-)-Cinchonidine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (8α,9R)-Cinchonan-9-ol
C0347-25G C0347-100G
TCIAC0347-25GEA 655 SEK
TCIAC0347-25G TCIAC0347-100G
(-)-Cinchonidine ≥98.0%
(-)-Cinchonidine
Formel: C₁₉H₂₂N₂O
MW: 294,4 g/mol
Kokpunkt: 465 °C (1013 hPa)
Smältpunkt: 199…202 °C
Densitet: 1,22 g/cm³ (20 °C)
MDL Number: MFCD00006783
CAS nummer: 485-71-2
EINECS: 207-622-3
Merck Index: 14,02286
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d47e7949f8fff486ed24cbeb76dd5cc44cf9bc3b&vid=d47e7949f8fff486ed24cbeb76dd5cc44cf9bc3b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -105 to -110 deg (C=1, EtOH)