7-Chloroquinaldine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 7-Chloro-2-methylquinoline

C1453-25G C1453-5G
TCIAC1453-25GEA 2100 SEK
TCIAC1453-25G TCIAC1453-5G
7-Chloroquinaldine ≥98.0%
7-Chloroquinaldine
Formel: C₁₀H₈ClN
MW: 177,63 g/mol
Kokpunkt: 87°C
MDL Number: MFCD00075436
CAS nummer: 4965-33-7
EINECS: 440-600-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db5fe812120177afde25f25e236bb6fe1fee5a7&vid=5db5fe812120177afde25f25e236bb6fe1fee5a7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR