1,2,4-Triazol-3-ylamin ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: Amitrol

A0432-25G A0432-500G
TCIAA0432-25GEA 215 SEK
TCIAA0432-25G TCIAA0432-500G
1,2,4-Triazol-3-ylamin ≥98.0% (by GC, titration analysis)
1,2,4-Triazol-3-ylamin
Formel: C₂H₄N₄
MW: 84,08 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 149…152 °C
Densitet: 1,138 g/cm³ (20 °C)
MDL Number: MFCD00005230
CAS nummer: 61-82-5
EINECS: 200-521-5
UN: 3077
ADR: 9,III
Merck Index: 14,00489
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22c48850dbac6c10c781d3f407803e88b9f113ff&vid=22c48850dbac6c10c781d3f407803e88b9f113ff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 154.0 to 159.0 deg-C
Solubility in Methanol faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR