1,2,3-Triazol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1,2,3-1H-Triazole, 1H-1,2,3-Triazole
T1880-25G
T1880-5G
TCIAT1880-25GEA
3450
SEK
TCIAT1880-25G
TCIAT1880-5G
1,2,3-Triazol ≥98.0%
1,2,3-Triazole
Formel:
C₂H₃N₃ MW: 69,07 g/mol Kokpunkt: 208 °C (1013 hPa) Smältpunkt: 23…25 °C Densitet: 1,192 g/cm³ (20 °C) |
MDL Number:
MFCD00014490 CAS nummer: 288-36-8 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow to Light orange powder to lump to clear liquid |
Purity (Nonaqueous Titration) | min. 98.0 % |
Freezing point | 21.0 to 24.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...