4-Bromophthalic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

B2257-1G B2257-25G B2257-5G
TCIAB2257-1GEA 215 SEK
TCIAB2257-1G TCIAB2257-25G TCIAB2257-5G
4-Bromophthalic acid ≥98.0%
4-Bromophthalic acid
Formel: C₈H₅BrO₄
MW: 245,03 g/mol
MDL Number: MFCD00043538
CAS nummer: 6968-28-1
EINECS: 230-183-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e0e8951fb729707da87dde2ab33685bf2f68a87&vid=0e0e8951fb729707da87dde2ab33685bf2f68a87&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR