Trimethyl orthoacetate 98,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

O0115-25ML O0115-500ML
TCIAO0115-25MLEA 215 SEK
TCIAO0115-25ML TCIAO0115-500ML
Trimethyl orthoacetate 98,0 GC_ANALYSMETOD
Trimethyl orthoacetate
Formel: C₅H₁₂O₃
MW: 120.15 g/mol
Kokpunkt: 108°C
Smältpunkt: -58
Flampunkt: 16°C
MDL Number: MFCD00008477
CAS nummer: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77195596c50ae3fd6535688b726a66f1455d39de&vid=77195596c50ae3fd6535688b726a66f1455d39de&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR