4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Cyano-5-fluorobenzotrifluoride
F0309-1G F0309-5G
TCIAF0309-1GEA 405 SEK
TCIAF0309-1G TCIAF0309-5G
4-Fluoro-2-(trifluoromethyl)benzonitrile ≥98.0%
4-Fluoro-2-(trifluoromethyl)benzonitrile
Formel: C₈H₃F₄N
MW: 189,11 g/mol
Kokpunkt: 52 - 55 °C (15 -rr)
Smältpunkt: 43…45 °C
Densitet: 1,35 g/cm³ (20 °C)
MDL Number: MFCD00061283
CAS nummer: 194853-86-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6b0bec4c4961fa88145a5186945ee95918fc555&vid=e6b0bec4c4961fa88145a5186945ee95918fc555&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 48.0 deg-C
Solubility in Methanol within almost transparency