4-Chloro-3-(trifluoromethyl)benzonitrile ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2-Chloro-5-cyanobenzotrifluoride

C2246-25G C2246-5G
TCIAC2246-25GEA 2675 SEK
TCIAC2246-25G TCIAC2246-5G
4-Chloro-3-(trifluoromethyl)benzonitrile ≥98.0%
4-Chloro-3-(trifluoromethyl)benzonitrile
Formel: C₈H₃ClF₃N
Kokpunkt: 120 - 125 °C (20 -rr)
Smältpunkt: 66…68 °C
MDL Number: MFCD00190117
CAS nummer: 1735-54-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8debb4e38c180f9533cd46f4c691abf6e3d9e55b&vid=8debb4e38c180f9533cd46f4c691abf6e3d9e55b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR