Dimethoxy(diphenyl)silane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Dimethoxydiphenylsilane, Diphenyldimethoxysilane

D1731-25ML D1731-500ML D1731-100ML
TCIAD1731-25MLEA 310 SEK
TCIAD1731-25ML TCIAD1731-500ML TCIAD1731-100ML
Dimethoxy(diphenyl)silane ≥98.0%
Dimethoxy(diphenyl)silane
Formel: C₁₄H₁₆O₂Si
MW: 244,37 g/mol
Kokpunkt: 110°C
Flampunkt: 157°C
MDL Number: MFCD00025690
CAS nummer: 6843-66-9
EINECS: 229-929-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be0d2070a6ef51ea5addee94fa5cef5302e6dc3c&vid=be0d2070a6ef51ea5addee94fa5cef5302e6dc3c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR