Dimethoxy(dimethyl)silane ≥98.0%

Leverantör: TCI
Fara

Synonyms: Dimethyldimethoxysilane

D1052-25ML D1052-500ML
TCIAD1052-25MLEA 215 SEK
TCIAD1052-25ML TCIAD1052-500ML
Dimethoxy(dimethyl)silane ≥98.0%
Dimethoxy(dimethyl)silane
Formel: C₄H₁₂O₂Si
MW: 120.22 g/mol
Kokpunkt: 81°C
Smältpunkt: -80
Flampunkt: –5°C
MDL Number: MFCD00025691
CAS nummer: 1112-39-6
EINECS: 214-189-4
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=065a43073b72eef6705bc23846e897d2eb01d55f&vid=065a43073b72eef6705bc23846e897d2eb01d55f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR