Diethoxy(diphenyl)silane ≥98.0%
Leverantör: TCI
Varning
Synonyms: Diphenyldiethoxysilane, Diethoxydiphenylsilane
D3067-100G D3067-25G
TCIAD3067-100GEA 2300 SEK
TCIAD3067-100G TCIAD3067-25G
Diethoxy(diphenyl)silane ≥98.0%
Diethoxy(diphenyl)silane
Formel: C₁₆H₂₀O₂Si
MW: 272.42 g/mol
Kokpunkt: 218°C
Flampunkt: 175°C
MDL Number: MFCD00015126
CAS nummer: 2553-19-7
EINECS: 219-860-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f9632718de51a3f275be7e13dd488d818dcc6a&vid=73f9632718de51a3f275be7e13dd488d818dcc6a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %