Diethoxy(dimethyl)silane ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Dimethyldiethoxysilane

D0454-25ML D0454-500ML D0454-100ML
TCIAD0454-25MLEA 230 SEK
TCIAD0454-25ML TCIAD0454-500ML TCIAD0454-100ML
Diethoxy(dimethyl)silane ≥98.0%
Diethoxy(dimethyl)silane
Formel: C₆H₁₆O₂Si
MW: 148,28 g/mol
Kokpunkt: 114°C
Smältpunkt: –87 °C
Densitet: 0,853 g/cm³ (20 °C)
Flampunkt: 15°C
MDL Number: MFCD00009068
CAS nummer: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=467a41ff9172774a082a1a6cdb84a81f8ac2963b&vid=467a41ff9172774a082a1a6cdb84a81f8ac2963b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR