Sudan IV
Leverantör: TCI EUROPE
Varning
Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol
O0100-25G O0100-100G
TCIAO0100-25GEA 215 SEK
TCIAO0100-25G TCIAO0100-100G
Sudan IV
Sudan IV
Formel: C₂₄H₂₀N₄O
MW: 380,45 g/mol
Smältpunkt: 188 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003893
CAS nummer: 85-83-6
EINECS: 201-635-8
Merck Index: 14,08393
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=208ba432e564eb0b71084ff79b1e9d2229454f3e&vid=208ba432e564eb0b71084ff79b1e9d2229454f3e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Orange to Amber to Dark red powder to crystal
Lambda max. 514.0 to 517.0 nm (THF)
Absorbance min. 0.780 (10mg/L, THF, 514.0 to 517.0 nm) (calcd.on dried substance)
Drying loss max. 2.5 %