1,1,3,3-Tetramethylbutylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2,4,4-Trimethylpentan-2-amine, 2-Amino-2,4,4-trimethylpentane, tert-Octylamine

T0143-25ML T0143-500ML
TCIAT0143-25MLEA 215 SEK
TCIAT0143-25ML TCIAT0143-500ML
1,1,3,3-Tetramethylbutylamine ≥98.0% (by GC, titration analysis)
1,1,3,3-Tetramethylbutylamine
Formel: C₈H₁₉N
MW: 129,25 g/mol
Kokpunkt: 140°C
Smältpunkt: –8 °C
Densitet: 0,805 g/cm³ (20 °C)
Flampunkt: 32°C
MDL Number: MFCD00008053
CAS nummer: 107-45-9
EINECS: 203-491-1
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb5fcef3781df39b18523050cc651f81685e025&vid=acb5fcef3781df39b18523050cc651f81685e025&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR