N,N,2,4,6-Pentamethylaniline ≥97.0%

Leverantör: TCI
Varning

P1767-25G P1767-5G
TCIAP1767-25GEA 1675 SEK
TCIAP1767-25G TCIAP1767-5G
N,N,2,4,6-Pentamethylaniline ≥97.0%
N,N,2,4,6-Pentamethylaniline
Formel: C₁₁H₁₇N
Kokpunkt: 215°C
Flampunkt: 79°C
MDL Number: MFCD00010596
CAS nummer: 13021-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a1761710aaa66325391208ddc40f79cb3134eaf&vid=5a1761710aaa66325391208ddc40f79cb3134eaf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR