Nuclear Fast Red
Leverantör: TCI EUROPE
Varning
Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
N0184-1G N0184-10G
TCIAN0184-1GEA 495 SEK
TCIAN0184-1G TCIAN0184-10G
Nuclear Fast Red
Nuclear Fast Red
Formel: C₁₄H₈NNaO₇S
MW: 357,28 g/mol
Smältpunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078493
CAS nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=617e4ac0bcbe17940fbcd795e088c2e06b3a2aa2&vid=617e4ac0bcbe17940fbcd795e088c2e06b3a2aa2&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Dark red to Brown powder to crystal
Lambda max. 532.0 to 536.0 nm (0.1mol/L NaOH sol.)
Absorbance (E1%1cm) min. 380 (0.1mol/L NaOH sol., 532.0 to 536.0 nm)