Natriumdiklorisocyanurat ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Sodium dichloroisocyanurate, Dichloro-1,3,5-triazinetrione sodium salt, Dichloroisocyanuric acid sodium salt

D1003-25G D1003-500G
TCIAD1003-25GEA 215 SEK
TCIAD1003-25G TCIAD1003-500G
Natriumdiklorisocyanurat ≥97.0%
Natriumdiklorisocyanurat
Formel: C₃Cl₂N₃NaO₃
MW: 219,95 g/mol
Smältpunkt: 225 °C
Densitet: 0,96 g/cm³ (20 °C)
MDL Number: MFCD00006036
CAS nummer: 2893-78-9
EINECS: 220-767-7
UN: 2465
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a433fd80d9274162ded82e53eb624c5c22c751&vid=a7a433fd80d9274162ded82e53eb624c5c22c751&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (Iodometric Titration) min. 97.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR